missing translation for 'onlineSavingsMsg'
Learn More

CD14 Antibody (LPSR/927), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-11438PCP

Product Code. 18610067

  • € 728.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD14
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Flow Cytometry
LPSR/927
Flow Cytometry
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Recombinant human CD14 protein (Uniprot: P08571)
0.1 mL
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
929
Store at 4°C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.