missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (RPA-M1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27215AF532
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD14 | |
| Human PHA-stimulated PBMC were used as immunogen. | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| RPA-M1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| 0.1 mL | |
| 929 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction