missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (RPA-M1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27215MFV610
This item is not returnable.
View return policy
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry
Specifications
| CD14 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| RPA-M1 | |
| Flow Cytometry | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Human PHA-stimulated PBMC were used as immunogen. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction