missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (TUK4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62994AF700
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product recognises the human CD14 cell surface antigen, expressed strongly by most peripheral blood monocytes and weakly on granulocytes. Clone Tuk4 has been shown to block SDF-induced chemotaxis of U937 cells in a dose-dependent manner(6). | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA | |
| TUK4 | |
| Flow Cytometry, ELISA | |
| P08571 | |
| CD14 | |
| Made to Human CD14 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction