missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD14 Monoclonal Antibody (TuK4), FITC

Mouse Monoclonal Antibody
€ 324.00 - € 1036.00
Specifications
| Antigen | CD14 |
|---|---|
| Clone | TuK4 |
| Concentration | 0.20mg/mL |
| Content And Storage | 4°C, store in dark |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10682774
![]() |
Invitrogen™
MHCD1401 |
500 μL |
€ 324.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10363293
|
Invitrogen™
MHCD1401-4 |
2 mL |
€ 1036.00
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.Specifications
| CD14 | |
| 0.20mg/mL | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| ASR | |
| PBS with 4mg/mL BSA and 0.1% sodium azide | |
| P08571 | |
| 929 | |
| Human CD14. | |
| Antibody |
| TuK4 | |
| 4°C, store in dark | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human | |
| Cd14 | |
| CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
| Cd14 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title