Learn More
CD14 Monoclonal Antibody (61D3), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 102.00 - € 263.00
Specifications
Antigen | CD14 |
---|---|
Clone | 61D3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16340974
|
Affymetrix eBioscience
53-0149-41 |
25 Tests |
€ 102.00
Pack of 25 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937245
|
Affymetrix eBioscience
53-0149-42 |
100 Tests |
€ 263.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14. Applications Reported: This 61D3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 61D3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Cd14 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd14 |
61D3 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
P08571 | |
929 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |