Learn More
CD14 Mouse anti-Human, PE, Clone: UCHM-1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65056-100TESTS
Description
The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with thymocytes and peripheral blood lymphocytes from a remission aml patient of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
| CD14 | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| UCHM-1 | |
| PE | |
| 100 Tests | |
| 929 | |
| IgG2a |