Learn More
CD144 Mouse anti-Human, BUV395, Clone: 55-7H1, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743708
Description
The 55-7H1 antibody monoclonal antibody specifically binds to a calcium-independent epitope on Cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is also known as CD144 or VE-Cadherin. CD144 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD144 | |
| 55-7H1 | |
| Brilliant Ultraviolet 395 | |
| VE-cadherin, Cadherin-5, CDH5, Vascular endothelial cadherin | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.