Learn More
CD146 Mouse anti-Human, PE, Clone: P1H12, BD
Mouse Monoclonal Antibody
€ 79.95 - € 346.42
Specifications
| Antigen | CD146 |
|---|---|
| Clone | P1H12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15810689
|
BD Biosciences
561013 |
25 Tests |
€ 80.25 € 79.95 / Each Save € 0.30 0.37% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15826228
|
BD Biosciences
550315 |
100 Tests |
€ 347.00 € 346.42 / Each Save € 0.58 0.17% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The P1H12 monoclonal antibody specifically binds to CD146. CD146 is a 118 kDa transmembrane glycoprotein also known as MCAM, MUC18, or Mel-CAM. CD146 is a member of the immunoglobulin superfamily and is expressed by angioblasts and mesenchymal stems cells and is strongly expressed by blood vessel endothelium and smooth muscle. CD146 is also expressed by melanoma cells, intermediate trophoblasts and a subpopulation of activated T cells. The P1H12 monoclonal antibody has been reported to block endothelial cell adhesion that is observed very early in embryogenesis. It can be useful in the study of embryologic vasculogenesis. This antibody is suitable for immunohistochemical staining of acetone-fixed frozen tissue sections, immunoprecipitation and ELISA.
Specifications
| CD146 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| P1H12 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Umbilical Vein Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.