Learn More
CD146 Mouse, anti-Rabbit, CoraLite Plus 555, Clone: P1H12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-65181-100TESTS
Description
The CD146 antibody from Proteintech is a mouse monoclonal antibody generated with cultured human umbilical cells of human CD146. This antibody recognizes rabbit, canine, mouse, human antigen. The CD146 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 555.
Specifications
| CD146 | |
| P1H12 | |
| CoraLite Plus 555 | |
| Mouse | |
| Primary | |
| Rabbit, Canine, Mouse, Human | |
| 155 kDa | |
| IgG1 κ |
| Flow Cytometry | |
| 5 μL/Test | |
| CD146, MCAM, MUC18 | |
| 100 Tests | |
| 4162 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.