Learn More
CD146 Mouse anti-Rabbit, PE, Clone: P1H12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65181-100TESTS
Description
The CD146 antibody from Proteintech is a mouse monoclonal antibody generated with cultured human umbilical cells of human CD146. This antibody recognizes rabbit, canine, mouse, human antigen. The CD146 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
| CD146 | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Rabbit, Canine, Mouse, Human |
| Flow Cytometry | |
| P1H12 | |
| PE | |
| 100 Tests | |
| 4162 | |
| IgG1 |