missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD148 Syrian Hamster anti-Mouse, BB700, Clone: 8A-1, BD OptiBuild™
Syrian Hamster Monoclonal Antibody
Brand: BD Biosciences 746045
This item is not returnable.
View return policy
Description
CD148 Monoclonal antibody specifically detects CD148 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD148 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Syrian Hamster | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 8A-1 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction