missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD15/Lewis X Antibody (BRA-4F1), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12070MFV500
This item is not returnable.
View return policy
Description
CD15/Lewis X Monoclonal antibody specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry
Specifications
| CD15/Lewis X | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| BRA-4F1 | |
| Flow Cytometry | |
| CD 15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| This antibody was raised by immunzing mice with human CD15/Lewis X. | |
| 0.1 mL | |
| Cancer | |
| 2526 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction