missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD15 Mouse anti-Human, BUV737, Clone: W6D3, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741876
This item is not returnable.
View return policy
Description
CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow Cytometry
Specifications
| CD15 | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| 3-fucosyl-N-acetyllactosamine; 3-FAL | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| W6D3 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto