missing translation for 'onlineSavingsMsg'
Learn More

CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-1548-42

234.01 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.23750kg

Product Code. 15540467

  • € 351.00 / 100 tests
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.

CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Cynomolgus Monkey, Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD154 (CD40 Ligand)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P29965
CD40LG
Affinity chromatography
RUO
102138630, 959
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
24-31
PE-eFluor 610
CD40LG
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Mouse
100 Tests
Primary
Cynomolgus Monkey, Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.