Learn More
Invitrogen™ CD155 Monoclonal Antibody (M048)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547658
Description
Clone M048 mouse monoclonal antibody detects a 75 kDa* protein on SDS-PAGE Native or denatured immunoblots of human A549, NCI-H466, NCI-H1299, MDA-MB-231 and A431 carcinomas. The antibody detects CD155 in the membranes and cytoplasm in NCI-H446 cells after immunocytochemical labeling. The antibody works for western blot, immunoprecipitation, immunocytochemistry, and ELISA capture.
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CD155 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P15151 | |
| PVR | |
| Clone (M048) was generated from a recombinant protein that included the extracellular region of human CD155 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M048 | |
| Unconjugated | |
| PVR | |
| 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.