missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (300907), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25301E
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| CD155/PVR | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 300907 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Mouse myeloma cell line NS0-derived recombinant human CD155/PVR, Gly27-Asn343, Accession # AAH15542 | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 5817 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction