missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (S04-2C7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 444.00 - € 680.00
Specifications
| Antigen | CD155/PVR |
|---|---|
| Clone | S04-2C7 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662204
|
Novus Biologicals
NBP3-19998-100ul |
100 μg |
€ 680.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613884
|
Novus Biologicals
NBP3-19998-50ul |
50 μg |
€ 444.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD155/PVR | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5817 | |
| IgG | |
| Affinity purified |
| S04-2C7 | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Recombinant protein of human CD155/PVR (Uniprot # P15151) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title