missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD155 Recombinant Rat Monoclonal Antibody (3F1)
Rat Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548217
This item is not returnable.
View return policy
Description
Specificity: This antibody is specific for murine CD155.
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CD155 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q3U0X8 | |
| PVR | |
| This antibody was raised by immunising rats with recombinant murine CD155, consisting of the entire ectodomain expressed as a HIS-tagged protein. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| ELISA, Flow Cytometry | |
| 3F1 | |
| Unconjugated | |
| PVR | |
| 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
| Rat | |
| Protein A | |
| RUO | |
| 52118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction