Learn More
CD158b1, b2, j Mouse anti-Human, BUV661, Clone: DX27, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 750066
Product Code. 17201465
  • ONLINE EXCLUSIVE PRICE
    € 389.00
    € 388.12 / 50µg
    Save € 0.88
    0.23% Off
Description
CD158b1, b2, j Monoclonal antibody specifically detects CD158b1, b2, j in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD158b1, b2, j
Monoclonal
Brilliant Ultraviolet 661
NKAT2/NKAT5/NKAT6; KIR2DL3/KIR2DS2/KIR2DL2
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Flow Cytometry
DX27
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
RUO
Human
IgG2a κ