Learn More
CD159c (NKG2C) Mouse anti-Human, Brilliant Violet 480, Clone: 134591, BD OptiBuild™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 748168
Product Code. 17269804
  • ONLINE EXCLUSIVE PRICE
    € 560.00
    € 542.19 / 50µg
    Save € 17.81
    3% Off
Description
CD159c (NKG2C) Monoclonal antibody specifically detects CD159c (NKG2C) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD159c (NKG2C)
Monoclonal
0.2 mg/mL
Mouse
RUO
CD159c (NKG2C)
Refrigerate (2 to 8°C)
Flow Cytometry
134591
Brilliant Violet 480