missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD16-2 Recombinant Rabbit Monoclonal Antibody (012), PE

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541007
This item is not returnable.
View return policy
Description
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
FcgRIV, also known as CD16-2, is an intermediate-affinity activating receptor for IgG2a and IgG2b. CD16-2 is the mouse homolog of human FcgRIIIA. CD16-2 is a low-affinity IgE receptor for all allotypes and the ligation of FcgRIV by antigen-IgE immune complexes promotes macrophage-mediated phagocytosis and is involved in lung inflammation.
Specifications
| CD16-2 | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| A0A0B4J1G0 | |
| Fcgr4 | |
| Recombinant Mouse FCGR4 protein. | |
| 100 Tests | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 012 | |
| PE | |
| Fcgr4 | |
| 4833442P21Rik; CD16-2; Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc gammaRIV; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; FcgammaRIV; Fcgr3a; Fcgr4; FcgRIV; Fcrl3; low affinity immunoglobulin gamma Fc region receptor III-A; Low affinity immunoglobulin gamma Fc region receptor IV | |
| Rabbit | |
| Protein A | |
| RUO | |
| 246256 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction