missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16/CD32 Antibody (93), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-27946JF549
This item is not returnable.
View return policy
Description
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| CD16/CD32 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 93 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3 | |
| Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors) | |
| 0.25 mL | |
| Adaptive Immunity, Immunology | |
| 2214 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction