missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16/CD32 Rat anti-Mouse, BB700, Clone: Ab93 (also known as 93 or Antibody93), BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 751699
This item is not returnable.
View return policy
Description
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD16/CD32 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms CD16/CD32 BB700 AB93 50 μg | |
| Mouse Pre-B cells | |
| RUO | |
| CD16/CD32 | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| Ab93 (also known as 93 or Antibody93) | |
| Brilliant Blue 700 (BB700) | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering