Learn More
CD16/CD32 Rat anti-Mouse, Brilliant Violet 510, Clone: 190909, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 747950
This item is not returnable.
View return policy
Description
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD16/CD32 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CD16/CD32 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 190909 | |
| Brilliant Violet 510 | |
| 50 μg | |
| Primary | |
| Mouse | |
| IgG2b κ |