missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16/CD32 Rat anti-Mouse, BUV737, Clone: Ab93 (also known as 93 or Antibody93), BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 751697
This item is not returnable.
View return policy
Description
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD16/CD32 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms CD16/CD32 BUV737 AB93 50 μg | |
| Mouse Pre-B cells | |
| RUO | |
| CD16/CD32 | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| Ab93 (also known as 93 or Antibody93) | |
| Brilliant Ultraviolet 737 | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction