Learn More
CD16 / CD32 Rat anti-Mouse, FITC, Clone: 2.4G2, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech FITC-65080-100UG
Product Code. 16811196
  • € 86.52 / 100µg
Description
The CD16 / CD32 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD16 / CD32. This antibody recognizes mouse antigen. The CD16 / CD32 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
Specifications
CD16 / CD32
Monoclonal
0.5 mg/mL
Rat
Primary
Mouse
Flow Cytometry
2.4G2
FITC
100 μg
14130
IgG2b