Learn More
CD16 / CD32 Rat anti-Mouse, FITC, Clone: 2.4G2, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech FITC-65080-100UG
Description
The CD16 / CD32 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD16 / CD32. This antibody recognizes mouse antigen. The CD16 / CD32 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD16 / CD32 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| 2.4G2 | |
| FITC | |
| 100 μg | |
| 14130 | |
| IgG2b |