missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, Brilliant Violet 650, Clone: 3G8, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 563692

 View more versions of this product

Product Code. 15823009

  • ONLINE EXCLUSIVE PRICE
    € 454.00
    € 453.52 / Each
    Save € 0.48
    0.11% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 3G8 monoclonal antibody specifically binds to the 50-65 kDa transmembrane form of the IgG Fc Receptor (Fc γRIII), a human NK cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. The 3G8 antibody blocks the binding of soluble immune complexes to granulocytes. The 3G8 antibody is reported (Vossebeld et al ., 1997) to increase intracellular calcium levels in human neutrophils by interacting with both Fc γRIIa and Fc γRIIIb molecules. This antibody has also been reported to induce homotypic neutrophil aggregation.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD16
Monoclonal
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human polymorphonuclear leukocytes
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
3G8
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Mouse
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.