missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Antibody (023), Alexa Fluor™ 750 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90073AF750
This item is not returnable.
View return policy
Description
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for ELISA, ELISA
Specifications
| CD160 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD160 (NP_008984.1; Met1-Leu158). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction | |
| 11126 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 023 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction