Learn More
CD160 Antibody (688327), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6700W
Product Code. 18546335
  • € 708.00 / 0.10mL
Description
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD160
Monoclonal
DyLight 650
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
688327
Flow Cytometry, CyTOF-ready
BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55
Chinese hamster ovary cell line CHO-derived recombinant human CD160, Ile27-Ser159, Accession # O95971
0.1 mL
Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction
11126
Store at 4°C in the dark.
IgG2b