missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41234F
This item is not returnable.
View return policy
Description
CD160 Polyclonal antibody specifically detects CD160 in Human, Mouse samples. It is validated for ELISA, Immunofluorescence
Specifications
| CD160 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunofluorescence | |
| FITC | |
| BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
| Antibody was raised against a 17 amino acid peptide near the center of human CD160. The immunogen is located within amino acids 50 - 100 of CD160 . Amino Acid Squence: SPETSLKQLRLKRDPG | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction | |
| 11126 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction