missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809703
This item is not returnable.
View return policy
Description
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.Specifications
| CD160 | |
| Monoclonal | |
| Unconjugated | |
| CD160 | |
| BY55, NK1, NK28 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11126 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2G2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95971 | |
| CD160 | |
| Human recombit protein fragment corresponding to amino acids 1-160 of human CD160 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction