missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Monoclonal Antibody (OTI3F8), TrueMAB™, OriGene
Description
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CD160 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3F8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ZMAT4 |
| Gene Accession No. | Q9H898 |
| Gene Alias | AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?