Learn More
Invitrogen™ CD160 Recombinant Rabbit Monoclonal Antibody (275), APC

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546744
Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD160 (Met1-Leu158) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
Specifications
| CD160 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.5% BSA and 0.1% sodium azide | |
| O95971 | |
| CD160 | |
| Recombinant protein ecoding Human CD160 (Met1-Leu158). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 275 | |
| APC | |
| CD160 | |
| AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11126 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.