missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (008), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-06112H
This item is not returnable.
View return policy
Description
CD161 Monoclonal antibody specifically detects CD161 in Mouse samples. It is validated for ELISA
Specifications
| CD161 | |
| Monoclonal | |
| HRP | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD161 (P27811; Gln67-His227). | |
| 0.1 mL | |
| Innate Immunity, Signal Transduction | |
| 3820 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 008 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction