missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (008), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 275.00 - € 453.00
Specifications
| Antigen | CD161 |
|---|---|
| Clone | 008 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613888
|
Novus Biologicals
NBP3-06112-100ul |
100 μg |
€ 453.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662998
|
Novus Biologicals
NBP3-06112-50ul |
50 μg |
€ 275.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD161 Monoclonal antibody specifically detects CD161 in Mouse samples. It is validated for ELISASpecifications
| CD161 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD161 (Uniprot#: P27811; Gln67-His227). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 008 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 3820 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title