missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (10/78), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne FAB9766D
This item is not returnable.
View return policy
Description
CD161 Monoclonal antibody specifically detects CD161 in Rat samples. It is validated for Flow Cytometry
Specifications
| CD161 | |
| Monoclonal | |
| DyLight 350 | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| Purified splenic NK cells from the LEW rat strain | |
| 0.1 mL | |
| Innate Immunity, Signal Transduction | |
| 3820 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 10/78. | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction