Learn More
CD161 Antibody (14F1F11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-14845AF647
Product Code. 18137251
  • € 605.00 / 0.10mL
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD161
Monoclonal
Alexa Fluor 647
Q12918
KLRB1
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody.
0.1 mL
Signal Transduction
3820
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
14F1F11
Western Blot, Immunohistochemistry-Paraffin
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Mouse
Protein G purified
RUO
Primary
The antibody has not been tested in species samples other than Human.
Store at 4C in the dark.