missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (14F1F11), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-14845V
This item is not returnable.
View return policy
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD161 | |
| Monoclonal | |
| DyLight 405 | |
| Q12918 | |
| KLRB1 | |
| Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
| 0.1 mL | |
| Signal Transduction | |
| 3820 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 14F1F11 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The antibody has not been tested in species samples other than Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction