missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161/NK1.1 Antibody (PK-136), Allophycocyanin - Chimeric, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20164APC
This item is not returnable.
View return policy
Description
CD161/NK1.1 Monoclonal antibody specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD161/NK1.1 | |
| Monoclonal | |
| APC | |
| CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| The original antibody was generated by immunizing (C3H X BALB)F1 mice with spleen cells enriched for NK-1 cells and bone marrow cells from CE mice. | |
| 0.1 mL | |
| Innate Immunity, Signal Transduction | |
| 17059 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| PK-136 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction