missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD162 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523970
This item is not returnable.
View return policy
Description
CD162 Polyclonal Antibody for Western Blot, Flow
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.
Specifications
| CD162 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q14242 | |
| SELPLG | |
| KLH conjugated synthetic peptide between 379-405 amino acids from the C-terminal region of human SELPLG. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| SELPLG | |
| CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6404 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction