missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (EDHu-1), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-40686PECY7
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Monkey, Primate, Rhesus Macaque, Sheep samples. It is validated for Flow Cytometry.
Specifications
| CD163 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD163 | |
| Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
| 0.1 mL | |
| Primary | |
| NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| EDHu-1 | |
| Flow Cytometry | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein A purified | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 9332.0 | |
| Human, Pig, Bovine, Guinea Pig, Monkey, Primate, Rhesus Macaque, Sheep | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction