missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (GHI/61), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43341R
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CD163 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD163 | |
| The immunogen for this antibody was CD163. | |
| 0.1 mL | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 9332 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| GHI/61 | |
| Western Blot, Flow Cytometry | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction