missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (OTI2B12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71420FR
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD163 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| CD163 | |
| Full length recombinant protein of human CD163 (NP_004235) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Affinity Purified | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 9332.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction