missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163, Mouse anti-Rat, DyLight 405, Clone: ED2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-39099V-100UL
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD163 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| CD163 | |
| Rat Spleen cell homogenate. Spleen cells from immunised BALB/c mice were fused with cells of the SP2/0-Ag 14 mouse myeloma cell line. | |
| 100 μL | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 9332 | |
| Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| ED2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes the rat ED2 cell surface glycoprotein. | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction