missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163, Rabbit anti-Human, Clone: K20-T, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB1652.100uL
This item is not returnable.
View return policy
Specifications
| CD163 | |
| Monoclonal | |
| Unconjugated | |
| In 20 mM Tris-HCl, pH 8.0 (20 mg/mL BSA, 0.05% sodium azide) | |
| M130/MM130 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| K20-T | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user. | |
| CD163 molecule | |
| CD163 | |
| A synthetic peptide corresponding to N-terminus of human CD163. | |
| RUO | |
| 9332 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction