missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163L1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92253-0.1ml
This item is not returnable.
View return policy
Description
CD163L1 Polyclonal antibody specifically detects CD163L1 in Mouse samples. It is validated for Western Blot
Specifications
| CD163L1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-550 of human CD163L1 (NP_777601.2). DCLHQNDVSVICSDGADLELRLADGSNNCSGRVEVRIHEQWWTICDQNWKNEQALVVCKQLGCPFSVFGSRRAKPSNEARDIWINSISCTGNESALWDCTYDGKAKRTCFRRSDAGVICSDKADLDLRLVGAHSPCYGRLEVKYQGEWGTVCHDRWSTRNAAVVCKQLGCGKPMHVFGMTYFKEASGPIWLDDVSCIGNES | |
| 0.1 mL | |
| CD Markers, Cell Biology, Immunology | |
| 283316 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction