Learn More
CD163L1 Antibody (CD163L1/7971), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24128AF647
Artikelnummer. 18529677
  • € 679.00 / 0.10mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Description
Description
CD163L1 Monoclonal antibody specifically detects CD163L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD163L1
Monoclonal
Alexa Fluor 647
CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202
Recombinant fragment of human CD163L1 protein (exact sequence is proprietary)
0.1 mL
CD Markers, Cell Biology, Immunology
283316
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
CD163L1/7971