missing translation for 'onlineSavingsMsg'
Learn More

CD163L1 Antibody (CD163L1/7971), Novus Biologicals™

Mouse Monoclonal Antibody

€ 232.00 - € 462.00

Specifications

Antigen CD163L1
Clone CD163L1/7971
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627794
View Documents
Novus Biologicals
NBP3-23697-100ug
100 μg
€ 462.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643164
View Documents
Novus Biologicals
NBP3-23697-20ug
20 μg
€ 232.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD163L1 Monoclonal antibody specifically detects CD163L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

CD163L1
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
CD Markers, Cell Biology, Immunology
10 mM PBS with 0.05% BSA
283316
IgG
Protein A or G purified
CD163L1/7971
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202
Recombinant fragment of human CD163L1 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.