missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD164 Antibody (006), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90082V
This item is not returnable.
View return policy
Description
CD164 Monoclonal specifically detects CD164 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD164 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 6 | |
| Flow Cytometry | |
| CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD164 (Q04900-1; Met1-Asp162). | |
| 0.1 mL | |
| Cancer | |
| 8763 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction